THERMODYNAMICS.
Statistical Thermo-Dynamics of Polymer Solutions and Blends
Polymer-Polymer Interactions Based on Mean Field Approximations
Hydrogen Bonding Systems
Polymer Blends as Viewed Byanalog Calorimetry
Crystalline Polymer Blends
Fundamentals of Blends of Rigid Chain (Liquid Crystal) Polymers
Thermodynamics of Polyolefin Blends
CHARACTERIZATION.
Morphology Characterization by Microscopy Techniques
Viscoelastic Characterization of Polymer Blends .
Optical Characterization: Light Scattering, Birefringence, and Ellipsometry
Neutron Scattering and Polymer Blends
STRUCTURE FORMATION.
Formulation and Characterization of Thermoset/Thermoplastic Blends
Chemical Reactions in Blends Based on Condensation Polymers: Transreactions, Molecular and Morphological Characterization
Morphology and Properties of Blends Containing Block Copolymers
Factors Influencing the Morphology in Immiscible Polymer Blends in Melt Processing
Reactive Compatibilization
Processing Aids
MECHANICAL PROPERTIES AND FRACTURE RESISTANCE.
Quasi-Elastic Mechanical Properties
Application of Fracture Mechanics for Characterization of Toughness of Polymer Blends
Characterizing Toughness Using Standard Empirical Tests
Deformation Mechanisms in Rubber-Toughened Polymers.
Strengthening Polymer-Polymer Interfaces
Core-Shell Impact Modifiers
Toughening of Semi-Crystalline Thermoplastics
Toughening of Epoxies
Fatigue Crack Propagation in Polymer Blends
BLENDING FOR SPECIFIC PERFORMANCE.
Transmission and Reflection of Light in Multiphase Media
Thermo-Mechanical Performance of Polymer Blends
Barrier Materials by Blending
REINFORCED BLENDS.
Reinforced Polymer Blends
Liquid Crystalline Polymer Blends
Microfibrillar Reinforced Composites From Polymer Blends
ELASTOMERIC BLENDS.
Elastomer Blends .
Thermoplastic Vulcanizates
RECYCLING.
Recycling of Polymer Blends and Mixture