General description
Emphasizes the general practices and instrumentation involving TLC and high performance TLC as well as their applications based on compound types, while providing a sound understanding of the underlying theory necessary for optimizing these techniques. Details novel qualitative and quantitative densitometric experiments on organic dyes, lipids, antibiotics, pharmaceuticals, organic acids, insecticides, and more! This latest edition also includes over 1,150 up-to-date references, new drawings of chromatograms, a glossary of important terms, and a list of supply and equipment sources.