General description
Laboratory instruments produce homogeneous, uniform samples for a variety of applications:
• Particle reduction, emulsifications, and reaction enhancements in chemical, pharmaceutical, and food research
• Tissue homogenization for biological and medical research
• Alcohol production
• Heavy metals analysis in sludge samples
• Homogenization of marine samples for separating bacteria aggregates
Other Notes
The slender design is for stand operation. 19mm diam. dispersing tool included. Order disperser
stand and boss head clamp separately.
Legal Information
IKA is a registered trademark of IKA Works, Inc.
ULTRA-TURRAX is a registered trademark of IKA Works, Inc.