|
| Z507539 Aldrich |
Ace gas dispersion tube with fritted discdisc O.D. × thickness 25 mm × 2 mm, porosity 70-100 μm (B) |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |