General description
This edition is fully revised to reflect the current state off the field. Significant additions include ultramicroelectrodes, modified electrodes, and scanning probe methods. Many chapters have been modified and improved, including electrode kinetics, voltammetric methods, and mechanisms of coupled chemical reactions. Takes the reader from the most basic chemical and physical principles through fundamentals of thermodynamics, kinetics, and mass transfer, to a thorough treatment of all important experimental methods. Treats application of electrochemical methods to elucidation of reaction mechanisms; double layer structure and surface processes, and their effects on electrode processes are developed from first principles; other key features include a chapter on operational amplifier circuits and electrochemical instrumentation, unique coverage of spectrometric and photochemical experiments, and Laplace transform and digital simulation techniques.