|
| Z512729 Aldrich |
Serrated natural rubber septablue rubber, bottom O.D. 23 mm, fits neck I.D., 20.5 mm, ST/NS: 24/40 |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |