General description
Special Features:
• Rapid, Even Heating up to 250°C
• Heavy-gauge stainless steel basin has a flat bottom and vertical walls for maximum stability; extra tall profile allows ample room for flask immersion without oil sloshing; dust cover keeps oil clean when not in use
• Immersion heating element sealed in a grounded metal sheath for safety; will not burn out even if operated dry for an extended period of time; detachable power cord
• Bath disassembles, separating all electrical parts from the stainless steel basin for easy cleaning
• Integral, basin-stabilizing clamp with holes that support 1/8 in. or 1/4 in. O.D. thermocouple probes; clamp attaches to 1/2 to 5/8 in. diameter vertical lattice rod or ring stand
• Suitable for magnetic stirring
• CE approved design
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.