Preface.
Introduction.
Abbreviations, Acronyms, and Symbols.
Chapter 1: Physical and Chemical Solvent Properties.
Chapter 2: Method Optimization.
Chapter 3: Method Validation and Ongoing Performance Evaluation.
Chapter 4: Alcohols.
Chapter 5: Alkanes and Alkyl Aromatics.
Chapter 6: Chlorinated Alkanes and Chlorinated Benzenes.
Chapter 7: Ethers.
Chapter 8: Keotones and Esters.
Chapter 9: Nitriles and Nitrogenous Solvents.
Chapter 10: Water, Dimethyl Sulfoxide, and Common Acidic Modifiers.
References.
Index.