General description
The CIS Chemical Inventory System-Standard is a high-performance, relational database system for tracking chemicals, materials, and other laboratory supplies. With CIS, you can:
•Maintain a listing of all the chemicals and materials in your facility
•Keep track of where they are and how much of them you have
•Monitor who uses them and how much
•Generate reports listing chemicals by location, vendor, name, CAS® number, formula, etc.
•Quickly access hazard information during an emergency
The program contains extensive on-line help, and a complete tutorial covering all aspects of operation. The CIS Chemical Inventory System-Standard is a great choice for Windows® chemical database management, providing both ease of use, and powerful features for editing, search and retrieval, and network environments.
•Container based tracking allows multiple lots, as well as unlimited vendors and sizes
•Locate containers based on container id’s that can be system or user defined
•Disposal tracking maintains a permanent record of all disposed containers
•Weight tracking algorithms for reporting total material in standard units
•User defined properties and fields provide information storage flexibility
•Threshold ceiling and floor warning limits
•Flag fields and reporting for SARA, TIER-2, and Form-R materials
•Highly flexible security and fully customizable password support
Legal Information
CIS Chemical Inventory System is a registered trademark of Chem SW, Inc.