Compatibility
Minimum system requirements: Windows® 95, 98, NT, or 2000
General description
“Modeling, property calculation, analysis, and database maintenance suite”
MolSuite 2000 gives you molecular modeling with great graphics, physical property calculations, statistical analysis, and database development and management all in one complete package. Model building is easy in ChemSite® Pro which provides fast minimization and displaying of any molecule, even crystals, in a 3D environment. Structures are easily copied and pasted to other MolSuite 2000 programs. Paste your molecule into Molecular Modeling Pro™ and run semi-empirical quantum mechanics and calculate up to 70 properties. Molecular Analysis Pro™ will enable you to use statistical regression techniques used in the plotting of chemical data. Use MolSuite to develop and maintain a database or use MolSuite® DB, which contains over 24,000 organic molecules with chemical properties. MolSuite 2000 is a desktop chemist’s affordable integrated suite.
Includes: ChemSite Pro, Molecular Modeling Pro, Molecular Analysis Pro and MolSuite DB (see descriptions above).
Legal Information
Molsuite is a trademark of ChemSW, Inc.