General description
In addition to capillary gas chromatography and GC-MS, carbon-13 NMR spectroscopy provides an alternative method for essential oils analysis. The excellent visual spacing of the signals enables oil samples to be analyzed without preliminary separation of their components. Also, information relating to the molecular structure of the oil constituents can be ascertained from the measured chemical shifts. This edition clearly demonstrates the power of the technique in the characterization of essential oils, based on 60 sample oils chosen for their industrial importance together with the 188 carbon-13 NMR spectra of significant components. Supporting data are presented using capillary gas chromatography.