General description
Developed by Best to the same exacting standards of its N-DEX family of hand protection. N-DEX Free's patented accelerator-free formulation creates a luminous, fun, yet PPE-conscious, high-visibility green glove. Powder-free, ambidextrous.
• Free from accelerators common in all disposable nitrile gloves, which have been linked to workplace conditions like Type IV dermatitis
• Free from natural rubber latex and Type I allergic reactions linked to latex proteins
Legal Information
Best is a registered trademark of Best Manufacturing Co.
N-DEX is a registered trademark of Best Manufacturing Co.