General description
Baths with circulators mounted on a stainless steel bridge. Ready for temperature applications to an external loop circuit and/or simultaneous operation in the bath tank.
TopTech series
• PID 2 temperature control with drift compensation
• Bright Multi-Display (LED) with resolution to 0.1 °C
• Keypad setting for actual value, 2 setpoints, upper and lower warning functions with audible signal (interval tone)
• Online communication with RS232 interface
• High temperature cut-out, adjustable
• Low liquid level protection: ability to refill bath before shut down
• Absolute temperature calibration 3 point: Simple correction of temperature registered on the internal and external measuring point.
• Model MC has pump pressure control, electronically adjustable pump capacity
Legal Information
Julabo is a registered trademark of Julabo Labortechnik GmbH.