General description
For EPA vials.
For the concentration of microplate extractions and reactions in vials or tubes with heat blocks, these evaporator systems have a fine needle valve which controls the total flow to the equalizer manifold providing equal gas flow to each well. The stainless steel needle height is easily adjustable to enable most sizes and capacities of microplates.
• Heats to 100°C for even faster drying times
• Digital temperature control
• Accuracy: 0.25% of Span
• Gas flow: 1 to 100 mL/needle
• On/Off power switch
Legal Information
Glas-Col is a registered trademark of Glas-Col Laboratory Products.