General description
The Evolution One glove is lightly chlorinated for greater glove stability. Longer and thicker for added protection, the Evolution One glove is made to provide greater manual dexterity and tactile sensitivity. Plus, it's powder-free and protein rated.
• Polymer coating for easy, powder-free donning
• Longer and thicker than typical examination gloves for greater protection over the wrist and arm
• Textured for a secure grip in wet or dry conditions
• Premium, low-modulus natural rubber latex for a true fit and reduced hand fatigue
Legal Information
Evolution One is a registered trademark of Microflex Medical Corp.
Microflex is a registered trademark of Microflex Medical Corp.