General description
Soft and smooth, supple, multiple glass yarn insulation with an additional braided protective lead around the heating cable. This heating tape corresponds to many different demands in laboratories and technical colleges. Protective braid - Nickel.
• Nominal temperature: 450 °C
• Min. bending radius: 10 mm
• Thickness: 5.5 mm
• Breadth: 30 mm
• Connection flex 0.5 M
• With connections box
Other Notes
Important: Heating tapes must be used in combination with a matching temperature regulator. The nominal temperature shown on the type label must not be exceeded.