General description
Laboratory regulator with many applications. Electronic Outlet up to 10 A. Combination of a power control unit and a temperature regulator with digital set-point adjustment. With the electronic power control unit the rated current may be reduced from 100% to 0 and thus be adjusted to the individual power requirements. When the sensor connection or the sensor element itself is broken the device turns off immediately. A control lamp indicates the fault. With a clamp the regulating device may be mounted to a stand. The clamp can be fixed at the rear of the device. Linear voltage: For the connection of a recording device, digital display etc. the regulator has linearized voltage tap. The device has a diode plug socket. Case material coated steel/aluminum.
• Two-position-regulator control system: P-PD
• Switching power at 230 V: 10A/2300W
• Main switch: 2-pole, luminous
• Power control: triac
• Degree of accuracy: 1%
• Servo-amplifier: 0.5 - 5%
• Instantaneous value tap: 10 mV
• Power control: 0 - 100%
• Safety class: I
• System of protection: I P 30 (DIN 40 050)
• Schuko socket outlet