General description
A rotating distributor in the extractor vessel is driven by a magnetic stirring plate. The extractor solvent fed from the condenser to the distributor is centrifugally forced through the small holes in the distributor ring as fine droplets into the liquid to be extracted, producing optimum exchange of matter. The liquid to be extracted also rotates in the extractor. The process is continuous and extraction is performed much more quickly than in traditional equipment. The rotary distributor on the inlet tube has a magnetic agitating rod, pivot nipple, bearings, antislip lock and separating ring with retaining ring.
Legal Information
Normag is a registered trademark of Normag Labor- und Prozesstechnik GmbH.