General description
Al-Si alloy top plate gives quick response and even temperature
Choice of robust aluminum or chemically resistant ceramic tops
Advanced safety features:
• Flashing "hot" warning light to warn when top plate is too hot to touch
• Spillproof design which helps deflect spills away from the controls and the user
• Independent safety circuit to protect against overheating
Microprocessor control for accurate temperature setting
General purpose, reliable hotplates with compact A4 footprint. These hotplates have microprocessor control of temperature allowing a calibrated scale to be printed around the control dial in ºC rather than an arbitrary 1 - 10 scale.
The "Hot" warning light will flash whenever the plate temperature is above 50°C and will operate even when the hotplate is turned off and connected to the mains.
Legal Information
Stuart is a registered trademark of Bibby Sterlin Ltd.