General description
Choice of robust aluminum or chemically resistant ceramic tops
Advanced safety features:
• Flashing 'Hot' warning light
• Spillproof design
• Independent safety circuit to protect against overheating
Microprocessor control for accurate temperature setting
Stylish and economical general purpose stirrer hotplates designed with safety as well as performance in mind. The cast aluminum body is shaped for stability and helps deflect spills away from the controls and the user. The 'Hot' warning light will flash whenever the plate temperature is above 50°C even when the hotplate is turned off and connected to the mains. Independent safety circuit prevents overheating in the event of control failure and internal electronic components are protected against corrosion. Powerful magnets and motor give stirring speed up to 1500rpm and volumes up to 15 L.
Fitting for retort rod and supplied with 2 x 25mm PTFE coated stir bars.
Al-Si alloy hot plate gives quick response and even temperatures
Legal Information
Stuart is a registered trademark of Bibby Sterlin Ltd.