General description
Time-saving Whatman qualitative grades are offered in this convenient form, which have major advantages over flat circles:
• Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analysis
• Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
• Increased total loading capacity because more filter area is available
• Maintained flow rate because of the reduction in filter paper contact with funnel side and, of course, the self-supporting shape of the filter itself
• The prepleating does not significantly affect any of the technical data and the same figures may be used for the flat circles.
Legal Information
Whatman is a registered trademark of Whatman International Ltd.