General description
This direct-controlled and heatable magnetic stirrer has exceptionally powerful magnetic stirrer for a stirring volume from 10 ml to 20 litres, even suitable for viscous media. The advantages of the superior hybrid drive are especially emphasized at high speed.
Automatic start-up and quick stop functions allow safe acceleration and safe, fast stopping of the stirring bar preventing danger of glass beaker breakage. Direct-controlled stirring give even stir bar acceleration without jerking, even at low speed.
The fully isolated aluminium heating plate means fast and energy-saving heating with precise processor control and safety circuit for unsupervised operation.
• Powerful stirring of up to 20 litres
• Rapid heating up to +320 °C
• Powerful speed: 2,000 rpm
• Compact, flat design
• RS 232-interface, can be used in networking
• Direct connection for contact thermometer
• LED display