Features and Benefits
• Resistant to water, ethanol, formalin, xylene and other solvents
• Will not run on embedding cassettes, frosted or painted slides
• Can be removed with acetic acid, ethanol/xylene mixture or paraffin
General description
General Use: Histology, Cytology
Writing Surface: Plastic embedding cassettes and frosted end of microscope slides.
Resistant To: Water, ethanol, formalin, xylene and other solvents. Removes from nonporous surfaces with acetic acid, chloroform or ethanol/xylene mixture.
Ultra-permanent ink that resists solvents such as xylene, ethanol, acetone and formalin. Ideal for histology and cytology. Markings will not run on embedding cassettes, frosted or painted slides or labware through repeated washes and solutions. Markings can be removed with acetic acid, ethanol/xylene mixture or paraffin. Testing is advised before use on actual specimens. (black)