| Z651109 Aldrich |
Hirschmann® burettes, straight stopcock, Class B50 mL volume, accuracy: 0.075 mL, PTFE key, black Graduations |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |