General description
Made from DURAN®. Conformity certified with dated batch identification. Main point ring graduation, intermediate stopcock with PTFE key, lateral valve stopcock with PTFE spindle. Calibrated to deliver (TD, EX) according to DIN 12 700 T5. Supplied with 2L amber glass supply bottle as standard.
Hirschmann supplies class A and AS volumetric glassware with a batch sign and the indication of the production year as standard at no additional charge. Therefore these measuring instruments are suitable for the use in laboratories certified DIN EN ISO.
Legal Information
Duran is a registered trademark of Schott AG.
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.