General description
For any bath tanks up to 50 L. With bath attachment clamp for wall thickness to 26 mm. Immersion depth 16.5 cm reducible to 14.5 cm (sleeves are supplied with each unit). Uniform mixing of bath liquid and space-saving design. Use the Julabo pump set for external temperature applications.
TopTech series, models MB and MC:
• Bright LED-display for the actual value + set point, resolution 0.1°C
• Adjustable high temp cut off low level liquid protection
• PID 2 temperature control with drift compensation
• Pump pressure 0.12 bar
• Keypad setting for actual value, 2 setpoints, upper and lower warning, pump stage, calibration and program capabilities
• Online communication with RS232 interface
• Low liquid level protection: ability to refill bath before shut down
• Absolute temperature calibration 3 point: simple correction of temperature registered on the internal and external measuring point.
• Model MC has pump pressure control, electronically adjustable pump capacity
Legal Information
Julabo is a registered trademark of Julabo Labortechnik GmbH.