General description
• External temperature applications
• Built-in cooling coil
• Compact design
• Electronically adjustable pressure pump capacity 'PPC'
• PID 2 temperature control with drift compensation
• Bright Multi-Display (LED) with resolution to 0.1 °C
• Pump pressure 0.45 bar
• Keypad setting for actual value, 3 set points, upper and lower warning functions and audible signal (interval tone)
• Online communication with integrated RS232 interface
• High temperature cut-out, adjustable
• Low liquid level protection: complete shut-down with continuous tone
• Absolute Temperature Calibration, 3 point: Simple correction of temperature registered on the internal and external measuring point.
Legal Information
Julabo is a registered trademark of Julabo Labortechnik GmbH.