General description
Nitri-Care is the economical medical examination version of the N-DEX family. Made in ISO 9002 quality-certified facility, Nitri-Care is inspected to a quality level of 1.5AQL. Nitri-Care does not contain any proteins of natural rubber and features the proprietary non-allergenic polymeric coating that aids in wet-hand donning.
• Tested according to EN 455
• Ambidextrous
• Patented "low-modulus" feature: the glove becomes a second skin
• Nitri-Care provides three times the puncture resistance of natural rubber and vinyl gloves of comparable thickness (puncture resistance is > 0.45 Kg)
• Best protection against latex allergies, viruses and bacteria
Legal Information
Best is a registered trademark of Best Manufacturing Co.
Nitri-Care is a registered trademark of Best Manufacturing Co.