General description
Flexible, cost effective precision furnaces. These models utilize a resistance wire heating element which is wound around the outside of a ceramic work tube making it an integral part of the heating element. If the tube is required to contain an atmosphere or is likely to be contaminated by spillage, an additional work tube should be used. The thermocouple is located in a protective position between the outside of the work tube and the heating element, allowing the full work tube diameter to be used and protect the thermocouple from mechanical damage. Fixed element tube mounted horizontally on a control box.
Heat up time 7 mins.
Other Notes
For safety reasons furnaces supplied with cable connection for hard wire only.
Legal Information
Carbolite is a registered trademark of Barlowworld Scientific Ltd.