General description
• New design allows for sample cooling
• Superior, magnetic driveless technology
• True, maintenance free magnetic coil drive
• Smooth operation as a result of no motor and no mechanicals couplings
• Extremely low noise
• Drive unit perfect for clean room applications because there is no fan
• Range of viscosity from 1 to 5,000 mPa.s
• Delivery includes 3 level plates for heigt adjustment of cooling vessel
• 3 year warranty
Other Notes
Not supplied with stand shown in picture
Protection class: IP30