General description
Whatman Nytran N (moderately charged) nylon membrane is ideal for applications that require a lower charge. It is designed for Southern and Northern blotting as well as colony and plaque lifts and Dot-/Slot-Blots. Nytran N is compatible with isotopic and non-isotopic detection methods. Nytran N membrane allows for excellent signal-to-noise ratios. The membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. Nytran N membrane is a highly consistent membrane with uniform pore size and distribution.It is available in 0.2 µm and 0.45 µm pore sizes for optimal retention of oligos and larger DNA fragments.
Legal Information
Nytran is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Whatman International Ltd.