General description
The TurboBlotter system is a rapid downward blotting device for high resolution transfer of DNA and RNA. Each kit provides supplies to complete fine transfers. Alkaline DNA transfers are completed in around one hour, neutral DNA and RNA transfers take only three hours.
• Greater speed and target resolution
• Convenient pre-cut and pre-stacked blotting papers
• Requires no power or vacuum source
Each TurboBlotter kit contains a 21 x 26 cm transfer device, as well as the blotting paper and membranes for five transfers.
Legal Information
TurboBlotter is a trademark of Whatman International Ltd.
Whatman is a registered trademark of Whatman International Ltd.