General description
For those professionals who insist on comfortable hand protection, the Diamond Grip Plus glove is a natural. Microflex performance at competitive pricing. This glove is manufactured with integrated polymer technologies for powder-free donning and product consistency. Beaded-cuff and fully textured for a secure grip in wet or dry conditions.
• Flash clorination provides consistent performance and greater comfort and fit
• Textured surface provides better grip in both wet and dry conditions
• Low modulus latex allows greater manual dexterity
• Integrated polymer technology creates a smooth surface for easier donning in all conditions
Thickness:
Palm 0.14 mm 5.1 mil
Cuff 0.11 mm 3.9 mil
Finger 0.17 mm 6.3 mil
Legal Information
Diamond Grip Plus is a trademark of Microflex Medical Corp.
Microflex is a registered trademark of Microflex Medical Corp.