General description
FreeForm SE is today's most advanced "super-soft" nitrile formulation offering a latex-like fit and feel no other synthetic glove can touch. Unlike conventional nitrile gloves, which tend to fit poorly and constrict, causing hands to work harder and fatigue faster, FreeForm SE works with you, continuously conforming to the contours of your hands as you move. FreeForm SE 's elasticity allows for a full range-of-motion, helping to minimize unnecessary hand stress and strain that can cause musculoskeletal injury over time.
• 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
• Textured fingers for a secure grip in wet and dry conditions
• Polymer coated for quick and easy donning, even in wet conditions
• Tested at factory, and again at Microflex to ensure product quality
• Exclusive patented formulation, Patent No. 6,451,893
Thickness
Cuff 0.11 mm 3.9 mil
Palm 0.10 mm 3.1 mil
Finger 0.14 mm 5.1 mil
Legal Information
Microflex is a registered trademark of Microflex Medical Corp.