General description
Synetron Plus latex examination gloves are a strong barrier against many chemicals and biohazards, and its comfortable during extended periods of wear and is textured for a secure grip. The polymer coating ensures product consistency and easy donning.
• Polymer-coating system provides effective donning in wet, damp or dry conditions
• Extended cuff for increased protection
• Made from soft natural rubber latex for improved elasticity and greater comfort
• Reduced protein rating
Gloves comply with personal protective equipment directive 89/686/eec, complex design category; European medical device directive 93/42/EEC, CE class 1; type tested to EN 374-2, EN 374-3, EN 388 & EN 420, and also complies with en 455 parts 1, 2, & 3.
Thickness
Cuff 0.17 mm 6.7 mil
Palm 0.23 mm 9.1 mil
Finger 0.30 mm 11.8 mil
Legal Information
Synetron is a trademark of Microflex Medical Corp.
Microflex is a registered trademark of Microflex Medical Corp.