grade
for ion chromatography
total impurities
≤10 μg/kg acetate
≤10 μg/kg formiate
≤10 μg/kg glycolate
≤10 μg/kg oxalate
refractive index
n20/D 1.34(lit.)
conductivity
≤2 μS/cm
bp
100 °C(lit.)
density
1.000 g/mL at 3.98 °C(lit.)
anion traces
bromide (Br-): ≤1 μg/kg
chloride (Cl-): ≤1 μg/kg
fluoride (F-): ≤1 μg/kg
iodide (I-): ≤1 μg/kg
nitrate (NO3-): ≤1 μg/kg
nitrite (NO2-): ≤1 μg/kg
phosphate (PO4-): ≤1 μg/kg
sulfate (SO42-): ≤1 μg/kg
cation traces
Al: ≤0.005 mg/kg
Ba: ≤0.005 mg/kg
Bi: ≤0.005 mg/kg
Ca: ≤0.01 mg/kg
Cd: ≤0.005 mg/kg
Co: ≤0.005 mg/kg
Cr: ≤0.005 mg/kg
Cu: ≤0.005 mg/kg
Fe: ≤0.005 mg/kg
K: ≤0.01 mg/kg
Li: ≤0.005 mg/kg
Mg: ≤0.005 mg/kg
Mn: ≤0.005 mg/kg
Mo: ≤0.005 mg/kg
NH4+: ≤0.05 mg/kg
Na: ≤0.01 mg/kg
Ni: ≤0.005 mg/kg
Pb: ≤0.005 mg/kg
Sr: ≤0.005 mg/kg
Zn: ≤0.005 mg/kg