|
| 01898 Fluka |
Adenosine Deaminase from calf intestinal mucosaBioChemika, in 50% glycerol, 0.01 M KH2PO4, pH 6.0, solution (clear), pale yellow, 150-250 units/mg protein (~10 mg/ml) |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |