Kit component also available separately
PEG 400 20% − Calcium acetate 0.1M − HEPES pH 7; 100 mM solution,
BioChemika 39719
MPEG 550 20% – Tris pH 8; 100 mM solution,
BioChemika 55914
PEG 1000 20% – MES pH 6.5; 100 mM solution,
BioChemika 51574
PEG 1500 20% – MES pH 6.5; 100 mM solution,
BioChemika 42876
PEG 2000 15% – Sodium chloride 0.15M – Tris pH 8.5 solution,
BioChemika 65838
MPEG 2000 20% – Citrate pH 6.5; 100 mM solution,
BioChemika 39712
MPEG 2000 20% − Sodium chloride 0.2M − HEPES pH 7; 100 mM solution,
BioChemika 39717
MPEG 2000 20% – Lithium sulfate 0.1M – MES pH 6; 100 mM solution,
BioChemika 39714
PEG 3350 15% − Ammonium sulfate 0.1M − HEPES pH 7.5; 100 mM solution,
BioChemika 39716
PEG 3350 20% – Sodium chloride 0.1M – Tris pH 8; 100 mM solution,
BioChemika 39711
PEG 3350 20% – Magnesium chloride 0.1M – MOPS pH 7; 100 mM solution,
BioChemika 39715
PEG 3350 20% − Cacodylate pH 6.5; 100 mM Solution,
BioChemika 39713
PEG 4000 8% – MES pH 6; 100 mM solution,
BioChemika 55913
PEG 4000 12% – Citrate pH 5.5; 100 mM solution,
BioChemika 68978
PEG 4000 12% – Ammonium sulfate 0.1M – Citrate pH 5.5; 100 mM solution,
BioChemika 18863
PEG 4000 15% − Magnesium chloride 0.1M − Cacodylate pH 6; 100 mM solution,
BioChemika 02131
PEG 4000 15% – Calcium chloride 0.1M – MES pH 6.5; 100 mM solution,
BioChemika 18999
PEG 4000 15% − 2-Propanol 10% − HEPES pH 7; 100 mM solution,
BioChemika 28496
PEG 4000 15% – Sodium chloride 0.1M – Tris pH 8; 100 mM solution,
BioChemika 02929
PEG 4000 20% − HEPES pH 7; 100 mM solution,
BioChemika 08586
MPEG 5000 10% − 2-Propanol 10% –− Cacodylate pH 6 solution,
BioChemika 02721
MPEG 5000 20% − HEPES pH 7.5; 100 mM solution,
BioChemika 23414
PEG 6000 8% – MES pH 6; 100 mM solution,
BioChemika 12145
PEG 6000 15% – Sodium acetate pH 5; 100 mM solution,
BioChemika 07857
PEG 6000 10% − Sodium chloride 0.1M − HEPES pH 7; 100 mM solution
16886
PEG 6000 10% − Calcium acetate 0.1M − HEPES pH 7.5; 100 mM solution,
BioChemika 07645
PEG 6000 15% – Tris pH 8; 100 mM solution,
BioChemika 11130
PEG 8000 8% – MES pH 6; 100 mM solution,
BioChemika 04162
PEG 8000 12% – Citrate pH 5.5; 100 mM solution,
BioChemika 18097
PEG 8000 12% − Sodium chloride 0.1M − Cacodylate pH 6; 100 mM solution,
BioChemika 11414
PEG 8000 12% – Magnesium acetate 0.1M – MES pH 6.5; 100 mM solution,
BioChemika 07164
PEG 8000 12% − Potassium chloride 0.1M − HEPES pH 7; 100 mM solution,
BioChemika 16950
PEG 8000 12% − Ammonium sulfate 0.1M − HEPES pH 7.5; 100 mM solution,
BioChemika 16885
PEG 8000 12% – Tris pH 8; 100 mM solution,
BioChemika 41009
PEG 8000 12% – Magnesium acetate 0.1M – Tris pH 8.5; 100 mM solution,
BioChemika 40681
PEG 20000 8% – MES pH 6; 100 mM solution,
BioChemika 41697
PEG 20000 12% – Tris pH 8; 100 mM solution,
BioChemika 68614
PEG 20000 12% − HEPES pH 7; 100 mM solution,
BioChemika 11147
PEG 20000 12% − Calcium chloride 0.1M − Cacodylate pH 6.5; 100 mM solution,
BioChemika 30939
Ammonium sulfate 1M – MES pH 6; 100 mM solution,
BioChemika 14148
Ammonium sulfate 1.5M – Citrate pH 5.5; 100 mM solution,
BioChemika 18061
Ammonium sulfate 1.6M – Cacodylate 6.5; 100 mM solution,
BioChemika 30529
Ammonium sulfate 1.6M − HEPES pH 7.5; 100 mM solution,
BioChemika 16395
Ammonium sulfate 1.8M – Tris pH 8.5; 100 mM solution, BioChemika
Sodium chloride 1 M – Tris pH 8; 100 mM solution,
BioChemika 16487
Potassium sodium tartrate 1 M − HEPES pH 7; 100 mM solution,
BioChemika 28871
Lithium sulfate 1.3M – Tris pH 8; 100 mM solution,
BioChemika 11442
Disodium hydrogenphosphate 0.375M - Dipotassium-hydrogenphosphate 0.1875M - Tris pH 8, 100mM solution,
BioChemika 12183