Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
03984
Fluka
Canada balsam solution
BioChemika
, for microscopy
CAS Number
8007-47-4
MDL number
MFCD00132800
Expand/Collapse All
Price and Availability
Descriptions
Application
embedding medium for microscopy
Physical form
dissolved in xylene
Properties
grade
for microscopy
product line
BioChemika
density
0.990 g/mL at 20 °C
Safety
Hazard Codes
Xn
Risk Statements
10-20/21-38
Safety Statements
36/37
RIDADR
UN 1307 3/PG 3
WGK Germany
2
Flash Point(F)
79 °F
Flash Point(C)
26 °C
Related Categories
...
Microscopy Reagents
>
Immersion Oils
...
Hematology and Histology
>
Routine Histology Stains
Scroll Up
Scroll Down
Related Information
FT-IR Raman
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
03984 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.