Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
03987
Fluka
Paraffin wax
BioChemika
, for histology, chunks (large)
CAS Number
8002-74-2
MDL number
MFCD00132833
EG/EC Number
232-315-6
Expand/Collapse All
Price and Availability
Properties
grade
for histology
product line
BioChemika
form
chunks (large)
transition temp
solidification point 55-60 °C
References
reference
FT-IR
2
(1), 14:C /
FT-IR
2
(1), 14:D /
FT-IR
2
(1), 14:A /
FT-IR
2
(1), 14:B /
Sax
6
, 2116
Safety
WGK Germany
NWG
RTECS
RV0350000
Flash Point(F)
235 °F
Flash Point(C)
113 °C
Related Categories
...
Embedding and Mounting Media
>
Embedding Media
...
Hematology and Histology
>
Routine Histology Stains
Scroll Up
Scroll Down
Related Information
FT-IR Condensed Phase
FT-IR Raman
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
03987 (Fluka)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.