Other Notes
Sales restrictions may apply
Preparation Note
Initial fractionation by heat shock.
Application
Bovine serum albumin (BSA) makes up approximately 50% of the total protein in serum. Although it may be used as a carrier for hapten immunization in animals other than goats and sheep, it is more commonly used as a carrier for hapten in immunoassays. The small size of BSA, its abundance in serum, and the similarity between BSA and albumins from other species makes it less desirable for antibody production.