Other Notes
Sales restrictions may apply
In the preparation of a bromouridine conjugate in EIA using a monoclonal antibody against 5-bromo-2-deoxyuridine for the assessment of lymphoid cell proliferation1
Preparation Note
from BSA by acetylation in order to inactivate nucleases.
Application
Bovine serum albumin (BSA) makes up approximately 50% of the total protein in serum. Although it may be used as a carrier for hapten immunization in animals other than goats and sheep, it is more commonly used as a carrier for hapten in immunoassays. The small size of BSA, its abundance in serum, and the similarity between BSA and albumins from other species makes it less desirable for antibody production.