Features and Benefits
Non-toxic Reagents Systems
With conventional electrophoresis reagents, there is always a risk to health. By contrast, the reagents in the ATX reagent system are non-toxic, environmentally friendly. Conventional reagents contain various toxic substances: buffer solutions contain barbituric acid, classified as a narcotic according to the Dangerous Drugs Act; staining solutions contain methanol and acetic acid which irritate the skin and airways; clarifying solutions contain methanol and dioxan isobutanol, a carcinogenic, inflammable substance which is classified as hazardous waste.
Stop using these toxic substances. For your electrophoresis, start using the non-toxic ATX reagent system and protect yourself and the environment against permanent damage.
The advantages are clear. ATX reagents are:
• non-toxic and non-irritant to skin and airways
• environmentally friendly
• disposable with regular waste water
• odorless
• non-inflammable
• stable at room temperature
• specially coordinated with ATX micro-solid strips
• available as ready-to-use solutions or as concentrates
• guaranteed to give excellent, reproducible results
Packaging
6 bottles for 1 L ready-to-use-solution