Components
Composition:
05497 Albumin Blue 580 Potassium salt Solution in 2-propanol, 10 mL
79438 Buffer for the determination of Albumine, 2×250 mL
05418 Albumin from human serum (HSA), 2×1 g
05476 Albumin from bovine serum (BSA), 10 g
09761 Calibrator Diluent Solution, 100 ml
General description
The components can also be ordered separately under the Fluka product numbers indicated.
Other Notes
For the specific and sensitive spectrofluorometric determination of albumine (1-200 mg/l) in natural matrices, using Albumine Blue 5801