grade
for luminescence
product line
BioUltra
expl. lim.
27 %
concentration
~1 M NH3 in H2O (T)
total impurities
insoluble matter, passes filter test
density
0.9 g/mL at 25 °C(lit.)
anion traces
chloride (Cl-): ≤0.5 mg/kg
phosphate (PO4-): ≤0.5 mg/kg
sulfate (SO42-): ≤2 mg/kg
cation traces
Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg
λ
neat
UV absorption
λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01
suitability
in accordance for luminescence test