grade
for trace analysis
TraceSELECT®
expl. lim.
27 %
concentration
≥25% NH3 in H2O (T)
density
0.91 g/mL at 20 °C
0.9 g/mL at 25 °C(lit.)
anion traces
bromide (Br-): ≤0.05 mg/kg
carbonate (CO2): ≤10 mg/kg
chloride (Cl-): ≤0.5 mg/kg
fluoride (F-): ≤0.1 mg/kg
iodide (I-): ≤0.05 mg/kg
nitrate (NO3-): ≤0.5 mg/kg
nitrite (NO2-): ≤0.1 mg/kg
phosphate (PO4-): ≤0.05 mg/kg
sulfate (SO42-): ≤0.1 mg/kg
cation traces
Ag: ≤0.001 mg/kg
Al: ≤0.005 mg/kg
As: ≤0.005 mg/kg
Au: ≤0.001 mg/kg
Ba: ≤0.001 mg/kg
Be: ≤0.001 mg/kg
Bi: ≤0.001 mg/kg
Ca: ≤0.01 mg/kg
Cd: ≤0.001 mg/kg
Co: ≤0.001 mg/kg
Cr: ≤0.001 mg/kg
Cs: ≤0.001 mg/kg
Cu: ≤0.001 mg/kg
Fe: ≤0.002 mg/kg
Ga: ≤0.001 mg/kg
Hg: ≤0.005 mg/kg
In: ≤0.001 mg/kg
K: ≤0.005 mg/kg
Li: ≤0.001 mg/kg
Mg: ≤0.001 mg/kg
Mn: ≤0.001 mg/kg
Mo: ≤0.001 mg/kg
Na: ≤0.01 mg/kg
Ni: ≤0.001 mg/kg
Pb: ≤0.001 mg/kg
Pt: ≤0.001 mg/kg
Rb: ≤0.001 mg/kg
Sn: ≤0.001 mg/kg
Sr: ≤0.001 mg/kg
Ti: ≤0.001 mg/kg
Tl: ≤0.001 mg/kg
V: ≤0.001 mg/kg
Zn: ≤0.001 mg/kg
Zr: ≤0.001 mg/kg