General description
Kit contains 20 peptones (10 g each)
18332 Peptone (vegetable)
51841 Peptone (vegetable) acid hydrolysate
19942 Peptone (vegetable), no 1
61854 Peptone (vegetable), no 2
92976 Peptone special (vegetable)
29185 Proteose Peptone (vegetable)
16922 Tryptone (vegetable)
12331 Tryptose (vegetable)
05138 Vegetable Extract
04316 Vegetable Extract no 1
49869 Vegetable Extract no 2
07436 Vegetable hydrolysate no 2
67381 Vegetable Infusion powder
95757 Vegetable Special Infusion powder
83059 Peptone from potatoes
93491 Peptone from broad bean
93492 Peptone from wheat
90765 Peptone from soybean, enzymatic digest
70178 Peptone from soybean, enzymatic digest
07022 Peptone from soybean, acid digest
Kit contains 20 peptones, extracts and hydrolysates from plant sources. This kit helps to evaluate the best growth properties of microganisms. With a peptide source specifically fitted to a microorganism, it is possible to increase the yield and improve the reproducibility of results.