Analysis Note
λmax. ~495 nm (MeOH)
Lucy-506 is provided as a 5000 × stock-solution in DMSO (5 mg/ml)
Application
Lucy-506 is a fluorescent stain for protein electrophoresis, with high sensitivity and easy, fast and robust staining procedure for all kinds of SDS gels. Protein staining by Lucy-506 does not interfere with subsequent MALDI-MS.
Sensitivity of routine procedure is ca. 3 ng/band, with linear response between 3 and 1000 ng/band
Legal Information
LUCY is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.