| 14454 Fluka |
2,2′-Bipyridylpuriss. p.a., redox indicator, for spectrophotometric det. of Fe, ≥99.0% (NT) |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |