Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
17119
Fluka
Aeromonas Selective Supplement
BioChemika
, for microbiology
Expand/Collapse All
Price and Availability
Descriptions
Components
Composition (per vial; sufficient for 500 mL medium)
Ampicillin 2.5 mg. Balanced with carbohydrates/buffer salts.
Application
An antibiotic supplement for the selective isolation of Aeromonas species.
Properties
grade
for microbiology
product line
BioChemika
shelf life
(limited shelf life, expiry date on the label)
storage temp.
2-8°C
Safety
Hazard Codes
Xn
Risk Statements
36/37/38-42/43
Safety Statements
22-26-36/37-45
WGK Germany
NWG
F
3
Related Categories
...
Media
>
Supplements
Scroll Up
Scroll Down
Related Information
17119 Data Sheet 88 kb
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
17119 (Fluka)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.